Chemical Identifiers
IUPAC Namedisodium (3Z)-5-hydroxy-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC16H10N2Na2O8S2
InChI KeyXTJONEUTTZZRAB-VZRGYXGKSA-L
SMILES[Na+].[Na+].OC1=CC(=CC2=C1C(=O)C(=NNC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol)468.36
View more