Thermo Scientific Chemicals

Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 192041000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 192041000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192041000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-17-8
IUPAC Name
dicaesium(1+) carbonate
Molecular Formula
CCs2O3
InChI Key
FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES
[Cs+].[Cs+].[O-]C([O-])=O
Appearance (Color)
White
Rubidium (Rb)
=<3000 ppm
Barium (Ba)
=<100 ppm
Appearance (Form)
Crystalline powder
Sodium (Na)
=<300 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text