Thermo Scientific Chemicals

Cesium iodide, 99.9%, for analysis, Thermo Scientific Chemicals

Catalog number: 192050250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium iodide, 99.9%, for analysis, Thermo Scientific Chemicals

Catalog number: 192050250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 192050250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-17-5
IUPAC Name
caesium(1+) iodide
Molecular Formula
CsI
InChI Key
XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES
[I-].[Cs+]
Purity
>=99.85 % (metals basis)
Rubidium (Rb)
=<50 ppm
Aluminium (Al)
=<10 ppm
Bismuth (Bi)
=<5 ppm
Nickel (Ni)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text