Thermo Scientific Chemicals

Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals

Catalog number: 192080025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals

Catalog number: 192080025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 192080025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-38-9
IUPAC Name
dichromium(3+) trioxidandiide
Molecular Formula
Cr2O3
InChI Key
UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Cr+3].[Cr+3]
Appearance (Color)
Green
Appearance (Form)
Powder
Assay
>=98.5 %
Loss on ignition
=<0.4 % (1000°C, 0.25h)
pH
5 to 7

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text