Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals
Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 192085000
Price (USD)/ Each
56.65
Online exclusive
66.80 
Save 10.15 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
66.80 
Save 10.15 (15%)
Add to cart
Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals
Catalog number192085000
Price (USD)/ Each
56.65
Online exclusive
66.80 
Save 10.15 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedichromium(3+) trioxidandiide
Molecular FormulaCr2O3
InChI KeyUOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Cr+3].[Cr+3]
Molecular Weight (g/mol)151.99
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Appearance (Form)Powder
Assay>=98.5 %
Loss on ignition=<0.4 % (1000°C, 0.25h)
pH5 to 7
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only