Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 192092500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Molecular Weight (g/mol)237.92
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to purple
Titration Complexometric98.0 to 102.0 %
Calcium (Ca)=<50 ppm
Magnesium (Mg)=<20 ppm
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only