Thermo Scientific Chemicals

Cobalt(II) carbonate hydrate, Thermo Scientific Chemicals

Catalog number: 192100500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cobalt(II) carbonate hydrate, Thermo Scientific Chemicals

Catalog number: 192100500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192100500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57454-67-8
IUPAC Name
λ²-cobalt(2+) carbonate
Molecular Formula
CCoO3
InChI Key
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
SMILES
[Co++].[O-]C([O-])=O
Titration Complexometric
>=45.5 % (Cobalt)
Cadmium (Cd)
=<50 ppm
Appearance (Form)
Powder
Insoluble matter
=<0.05 % (in HCl)
Copper (Cu)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text