Thermo Scientific Chemicals

Sodium tetrafluoroborate, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 192125000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium tetrafluoroborate, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 192125000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192125000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13755-29-8
IUPAC Name
sodium tetrafluoroboranuide
Molecular Formula
BF4Na
InChI Key
KGJZTOFHXCFQIV-UHFFFAOYSA-N
SMILES
[Na+].F[B-](F)(F)F
Titration
>=97.0 %
Appearance (Color)
White to light grey
Heavy metals (as Pb)
=<30 ppm
Chloride (Cl)
=<200 ppm
Sulfate (SO4)
=<1000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text