Thermo Scientific Chemicals

Chromium(III) acetylacetonate, 97%, Thermo Scientific Chemicals

Catalog number: 192131000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Chromium(III) acetylacetonate, 97%, Thermo Scientific Chemicals

Catalog number: 192131000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192131000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21679-31-2
IUPAC Name
chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21CrO6
InChI Key
JWORPXLMBPOPPU-LNTINUHCSA-K
SMILES
[Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Appearance (Color)
Purple
Elemental Analysis
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text