Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals
Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
500 g
Packaging:
Glass bottle
Catalog number 192150025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Color scale=<70 APHA
Hydroxyl value250 to 276 mg KOH/g
Water=<0.15 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only