Thermo Scientific Chemicals

Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals

Catalog number: 192155000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals

Catalog number: 192155000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 192155000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25322-69-4
Molecular Formula
(C3H6O)n
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
[H]OC(C)CO
MDL Number
MFCD00084446
Appearance (Form)
Clear viscous liquid
Color scale
=<70 APHA
Hydroxyl value
250 to 276 mg KOH/g
Water
=<0.15 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text