Poly(ethylene glycol), average M.W. 200
Poly(ethylene glycol), average M.W. 200
Poly(ethylene glycol), average M.W. 200
Poly(ethylene glycol), average M.W. 200
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 200

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic drum
Catalog number 192210051
Price (USD)
291.65
Special offer
Online exclusive
343.00
Save 51.35 (15%)
Each
-
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
291.65
Special offer
Online exclusive
343.00
Save 51.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Hydroxyl value534 to 591 mg KOH/g
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
Appearance (Form)Clear liquid
Water=<0.5 % (K.F.)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473719Certificate of AnalysisJun 23, 2025192210010, 192210051
A0465980Certificate of AnalysisSep 19, 2024192210010, 192210051
A0459672Certificate of AnalysisJan 26, 2024192210010, 192210051
A0455051Certificate of AnalysisAug 25, 2023192210010, 192210051
A0394420Certificate of AnalysisDec 15, 2022192210010, 192210051
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection