Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 192220025
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
Add to cart
Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals
Catalog number192220025
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
CAS25322-68-3
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
Hydroxyl value356 to 394 mg KOH/g
Water=<0.5 % (K.F.)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only