Poly(ethylene glycol), average M.W. 400, Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 400, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 400, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic drum
Catalog number 192230051
Price (USD)/ Each
334.65
Online exclusive
394.00 
Save 59.35 (15%)
-
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
334.65
Online exclusive
394.00 
Save 59.35 (15%)
Add to cart
Poly(ethylene glycol), average M.W. 400, Thermo Scientific Chemicals
Catalog number192230051
Price (USD)/ Each
334.65
Online exclusive
394.00 
Save 59.35 (15%)
-
Add to cart
Chemical Identifiers
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
CAS25322-68-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4650 to 1.4680 (20°C, 589 nm)
Water=<1.0 % (K.F.)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only