Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals

Catalog number: 192240010
1 L, Plastic bottle, Each
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals

Catalog number: 192240010
1 L, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192240010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Refractive index
1.4660 to 1.4700 (20°C, 589 nm)
Appearance (Color)
Clear colourless to almost colourless
Hydroxyl value
178 to 197 mg KOH/g
Appearance (Form)
Viscous liquid or low melting solid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text