Poly(ethylene glycol), average M.W. 1500, Thermo Scientific Chemicals
This product ships via ground from US northeast.
Poly(ethylene glycol), average M.W. 1500, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 1500, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 192260051
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
Add to cart
Poly(ethylene glycol), average M.W. 1500, Thermo Scientific Chemicals
Catalog number192260051
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Waxy flakes
Water=<1.0 % (K.F.)
Appearance (Color)White
Infrared spectrumConforms
Hydroxyl value70 to 80 mg KOH/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only