Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 6000, Thermo Scientific Chemicals

Catalog number: 192280010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 6000, Thermo Scientific Chemicals

Catalog number: 192280010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192280010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Water
=<0.5 % (K.F.)
Infrared spectrum
Conforms
Hydroxyl value
17 to 20 mg KOH/g
Appearance (Color)
White
Appearance (Form)
Powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text