Thermo Scientific Chemicals

Barium nitrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 192555000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Barium nitrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 192555000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 192555000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10022-31-8
IUPAC Name
barium(2+) dinitrate
Molecular Formula
BaN2O6
InChI Key
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Form)
Crystals
Chloride (Cl)
=<5 ppm
Strontium (Sr)
=<0.1 %
Appearance (Color)
White
Potassium (K)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text