Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Thermo Scientific Chemicals

Bismuth(III) oxide, 98+%, pure

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Catalog number 192620050
Price (USD)
-
Request bulk or custom format
Chemical Identifiers
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Yellow
Assay98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Share catalog number, name or link