Thermo Scientific Chemicals

Cadmium chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 192690025
2.5 kg, Each
Thermo Scientific Chemicals

Cadmium chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 192690025
2.5 kg, Each
Quantity
Catalog number: 192690025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10108-64-2
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Sulfate (SO4)
=<0.05 %
Appearance (Color)
White to off-white
Titration Argentometric
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text