Thermo Scientific Chemicals

Calcium chloride, 96%, for biochemistry, anhydrous, Thermo Scientific Chemicals

Catalog number: 192735000
500 g, Each
Thermo Scientific Chemicals

Calcium chloride, 96%, for biochemistry, anhydrous, Thermo Scientific Chemicals

Catalog number: 192735000
500 g, Each
Quantity
Catalog number: 192735000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-52-4
IUPAC Name
calcium dichloride
Molecular Formula
CaCl2
InChI Key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ca++]
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<5 ppm
Copper (Cu)
=<0.0005 %
Phosphate (PO4)
=<0.001 %
Sulfate (SO4)
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text