Thermo Scientific Chemicals

Cesium chloride, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 192812500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium chloride, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 192812500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 192812500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7647-17-8
IUPAC Name
caesium(1+) chloride
Molecular Formula
ClCs
InChI Key
AIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES
[Cl-].[Cs+]
Appearance (Form)
Powder
Appearance (Color)
White
Total trace metal impurities
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text