Thermo Scientific Chemicals

Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 192820500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 192820500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 192820500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-17-5
IUPAC Name
caesium(1+) iodide
Molecular Formula
CsI
InChI Key
XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES
[I-].[Cs+]
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Total trace metal impurities
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text