Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number 192870100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Cobalt (Co)21.0 to 23.0 %
Total trace metal impurities=<0.001 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only