Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
Catalog number 193070010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white to silver grey
Appearance (Form)Metallic solid or liquid
Total trace metal impurities=<1 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only