Thermo Scientific Chemicals

Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193070250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193070250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 193070250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-55-3
IUPAC Name
gallium
Molecular Formula
Ga
InChI Key
GYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES
[Ga]
Appearance (Color)
Silver white to silver grey
Appearance (Form)
Metallic solid or liquid
Total trace metal impurities
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text