Thermo Scientific Chemicals

Ammonium iodide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193210500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium iodide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193210500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193210500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12027-06-4
IUPAC Name
ammonium iodide
Molecular Formula
H4IN
InChI Key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES
[NH4+].[I-]
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or crystals
Appearance
May turn yellow on storage
Residue after ignition
=<0.05 %
Insoluble matter
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text