Lead(II) chloride, 99%, Thermo Scientific Chemicals
Lead(II) chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 193310500
Price (USD)/ Each
35.65
Online exclusive
39.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.80 
Save 4.15 (10%)
Add to cart
Lead(II) chloride, 99%, Thermo Scientific Chemicals
Catalog number193310500
Price (USD)/ Each
35.65
Online exclusive
39.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-95-4
IUPAC Nameλ²-lead(2+) dichloride
Molecular FormulaCl2Pb
InChI KeyHWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration Complexometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only