Lead(II) chloride, 99%
Lead(II) chloride, 99%
Lead(II) chloride, 99%
Lead(II) chloride, 99%
Thermo Scientific Chemicals

Lead(II) chloride, 99%

CAS: 7758-95-4 | Cl2Pb | 278.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Plastic bottle
Catalog number 193312500
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7758-95-4
IUPAC Nameλ²-lead(2+) dichloride
Molecular FormulaCl2Pb
InChI KeyHWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration Complexometric>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection