Thermo Scientific Chemicals

Lead(II) nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193320500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Lead(II) nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193320500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193320500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10099-74-8
IUPAC Name
λ²-lead(2+) dinitrate
Molecular Formula
N2O6Pb
InChI Key
RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total trace metal impurities
=<0.001 %
Appearance (Form)
Crystalline solid
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text