Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193395000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193395000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 193395000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-25-7
IUPAC Name
dilithium(1+) hydrate sulfate
Molecular Formula
H2Li2O5S
InChI Key
RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES
[Li+].[Li+].O.[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration after Ion exchange
>=99.0 % (dried basis) Li2SO4
Loss on drying
13.0 % to 15.0 % (1 g, 150°C)
Insoluble matter
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text