Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193455000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 193455000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193455000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-34-9
IUPAC Name
manganese(2+) tetrahydrate dichloride
Molecular Formula
Cl2H8MnO4
InChI Key
CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES
O.O.O.O.[Cl-].[Cl-].[Mn++]
Insoluble matter
=<0.005 %
Potassium (K)
=<0.01 %
Appearance (Form)
Crystals
pH
3.5 to 6.0 (5% soln. at 25°C)
Magnesium (Mg)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text