Thermo Scientific Chemicals

Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193470250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193470250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193470250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-13-9
IUPAC Name
dioxomanganese
Molecular Formula
MnO2
InChI Key
NUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILES
O=[Mn]=O
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
Grey to black
Total trace metal impurities
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text