Thermo Scientific Chemicals

Mercury(II) iodide, ACS reagent, Red, Thermo Scientific Chemicals

Catalog number: 193505000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Mercury(II) iodide, ACS reagent, Red, Thermo Scientific Chemicals

Catalog number: 193505000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 193505000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7774-29-0
IUPAC Name
mercury(2+) diiodide
Molecular Formula
HgI2
InChI Key
YFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES
[I-].[I-].[Hg++]
Appearance (Form)
Crystalline powder
Solubility
(In potassium iodide soln.) Passes test
Appearance (Color)
Red
Titration Iodometric
>=99.0 % (Dried basis)
Mercurous Mercury (as Hg)
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text