Thermo Scientific Chemicals

Mercury(II) oxide, 99+%, ACS reagent, red, Thermo Scientific Chemicals

Catalog number: 193510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Mercury(II) oxide, 99+%, ACS reagent, red, Thermo Scientific Chemicals

Catalog number: 193510250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193510250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
21908-53-2
IUPAC Name
oxomercury
Molecular Formula
HgO
InChI Key
UKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILES
O=[Hg]
Titration Complexometric
>=99.0 %
Insoluble matter
=<0.03 % (in dilute HCl)
Residue after reduction
=<0.025 %
Sulfate (SO4)
=<0.015 %
Chloride (Cl)
=<0.025 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text