Palladium(II) oxide, 99.995%, (trace metal basis)
Palladium(II) oxide, 99.995%, (trace metal basis)
Palladium(II) oxide, 99.995%, (trace metal basis)
Palladium(II) oxide, 99.995%, (trace metal basis)
Thermo Scientific Chemicals

Palladium(II) oxide, 99.995%, (trace metal basis)

CAS: 1314-08-5 | OPd | 122.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 193680050
Price (EUR)
1.164,50
Online Exclusive
1.370,00
Save 205,50 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
1.164,50
Online Exclusive
1.370,00
Save 205,50 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Namepalladium(2+) oxidandiide
Molecular FormulaOPd
InChI KeyJQPTYAILLJKUCY-UHFFFAOYSA-N
SMILES[O--].[Pd++]
Molecular Weight (g/mol)122.42
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granules and/or chunks
Assay85.2 to 88.7 % (Pd)
Total trace metal impurities=<50 ppm
Appearance (Color)Grey to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Share catalog number, name or link