Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 193690250
Price (USD)/ Each
5,870.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
5,870.00
Add to cart
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Catalog number193690250
Price (USD)/ Each
5,870.00
-
Add to cart
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.01 %
Appearance (Color)Grey to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only