Thermo Scientific Chemicals

cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193760010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 193760010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 193760010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15663-27-1
IUPAC Name
dichloroplatinumbis(ylium) diamine
Molecular Formula
Cl2H6N2Pt
InChI Key
BSJGASKRWFKGMV-UHFFFAOYSA-L
SMILES
N.N.Cl[Pt++]Cl
Infrared spectrum
Conforms
Assay
64.0 to 66.0 % (Pt)
Total trace metal impurities
=<0.01 %
Appearance (Color)
Yellow to orange
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text