cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number 193760050
Price (USD)/ Each
817.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
817.00
Add to cart
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals
Catalog number193760050
Price (USD)/ Each
817.00
-
Add to cart
Chemical Identifiers
CAS15663-27-1
IUPAC Namedichloroplatinumbis(ylium) diamine
Molecular FormulaCl2H6N2Pt
InChI KeyBSJGASKRWFKGMV-UHFFFAOYSA-L
SMILESN.N.Cl[Pt++]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay64.0 to 66.0 % (Pt)
Total trace metal impurities=<0.01 %
Appearance (Color)Yellow to orange
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only