Potassium chloride, extra pure
Potassium chloride, extra pure
Potassium chloride, extra pure
Potassium chloride, extra pure
Thermo Scientific Chemicals

Potassium chloride, extra pure

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 193780050
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=99.0 %
pH5.0 to 8.5 (5 % in water at 25°C)
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
I want to label Golgi bodies in my fixed cells. Is a dye-conjugated lectin like wheat germ agglutinin or concanavalin A a good choice?
Can I use CellMask Plasma membrane stains or Alexa Fluor dye labeled wheat germ agglutinin to label the plasma membrane of my paraffin sections?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471953Certificate of AnalysisMay 17, 2025193780050, 193780010
A0468652Certificate of AnalysisJan 31, 2025193780050, 193780010
A0464007Certificate of AnalysisJul 02, 2024193780050, 193780010
A0458894Certificate of AnalysisDec 20, 2023193780050, 193780010
A0454061Certificate of AnalysisAug 25, 2023193780050, 193780010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection