Potassium sulfate, pure, Thermo Scientific Chemicals
Potassium sulfate, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium sulfate, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 193810050
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
Add to cart
Potassium sulfate, pure, Thermo Scientific Chemicals
Catalog number193810050
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (130°C)
Appearance (Color)White
Titration with HClO4>=99.0 %
pH5.5 to 8.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only