1x1 image pixel for data collection
Potassium sulfate, pure
Potassium sulfate, pure
Potassium sulfate, pure
Potassium sulfate, pure
Thermo Scientific Chemicals

Potassium sulfate, pure

CAS: 7778-80-5 | K2O4S | 174.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 193810250
Price (USD)
549.00
Each
Add to cart
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (USD)
549.00
Each
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (130°C)
Appearance (Color)White
Titration with HClO4>=99.0 %
pH5.5 to 8.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link