Thermo Scientific Chemicals

Ammonium hexafluorosilicate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194000100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium hexafluorosilicate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194000100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194000100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16919-19-0
IUPAC Name
diammonium hexafluorosilanediuide
Molecular Formula
F6H8N2Si
InChI Key
ITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES
[NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
Total trace metal impurities
=<10 ppm
Appearance (Color)
White
Appearance (Form)
Crystalline powder and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text