Silver sulfate, ACS reagent, Thermo Scientific Chemicals
Silver sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver sulfate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 194070100
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Silver sulfate, ACS reagent, Thermo Scientific Chemicals
Catalog number194070100
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric>=98.0 %
Insoluble matter=<0.02 % (in water and AgCl)
Nitrate (NO3)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only