Silver sulfate, ACS reagent
Silver sulfate, ACS reagent
Silver sulfate, ACS reagent
Silver sulfate, ACS reagent
Thermo Scientific Chemicals

Silver sulfate, ACS reagent

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 194071000
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric>=98.0 %
Insoluble matter=<0.02 % (in water and AgCl)
Iron (Fe)=<0.001 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468377Certificate of AnalysisJan 31, 2025194070100, 194071000, 194075000
A0465294Certificate of AnalysisAug 08, 2024194070100, 194071000, 194075000
A0454850Certificate of AnalysisAug 25, 2023194070100, 194071000, 194075000
A0401143Certificate of AnalysisDec 15, 2022194070100, 194071000, 194075000
A0396875Certificate of AnalysisDec 14, 2022194070100, 194071000, 194075000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection