Thermo Scientific Chemicals

Strontium chloride hexahydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 194120010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Strontium chloride hexahydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 194120010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 194120010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-70-4
IUPAC Name
strontium(2+) hexahydrate dichloride
Molecular Formula
Cl2H12O6Sr
InChI Key
AMGRXJSJSONEEG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Appearance (Color)
White
Appearance (Form)
Crystals
Titration Complexometric
>=98.5 %
Barium (Ba)
=<0.1 %
Calcium (Ca)
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text