Tantalum(V) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Tantalum(V) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum(V) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 194170500
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
Add to cart
Tantalum(V) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Catalog number194170500
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-61-0
IUPAC Name[(dioxotantalio)oxy]dioxotantalum
Molecular FormulaO5Ta2
InChI KeyPBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILESO=[Ta](=O)O[Ta](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Total trace metal impurities=<0.01 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only