Chemical Identifiers
CAS14694-95-2
IUPAC Nameλ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular FormulaC54H45ClP3Rh
InChI KeyIXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more