Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 194610500
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
Add to cart
Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Catalog number194610500
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay>=99.99 % (Y2O3) TREO base
Total conc. of imp.as REO=<100 ppm
Loss on ignition=<1.0 % (1000°C) Material base
Particle size2 to 10 µm (d50)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only