Thermo Scientific Chemicals

Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194612500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194612500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194612500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-36-9
IUPAC Name
diyttrium(3+) trioxidandiide
Molecular Formula
O3Y2
InChI Key
RUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Y+3].[Y+3]
Appearance (Form)
Powder
Assay
>=99.99 % (Y2O3) TREO base
Total conc. of imp.as REO
=<100 ppm
Loss on ignition
=<1.0 % (1000°C) Material base
Particle size
2 to 10 µm (d50)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text